tert-Butyl-4-oxoazepane-1-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-BOC-Hexahydro-1H-azepin-4-oneTert-butyl 4-oxoazepane-1-carboxylate4-Oxoazepane-1-carboxylic acid tert-butyl ester

H55214-03 H55214-06 H55214-MD
AAH55214-06EA 472.99 USD
AAH55214-06 AAH55214-MD AAH55214-03
tert-Butyl-4-oxoazepane-1-carboxylate 98%
tert-Butyl-4-oxoazepane-1-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788435
CAS Number: 188975-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR