Bicyclohexane-4,4'-dione monoethylene ketal 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-(4-Oxocyclohexyl)-1,4-dioxaspiro[4.5]decane

H55069-14 H55069-06
AAH55069-06EA 42.36 USD
AAH55069-06 AAH55069-14
Bicyclohexane-4,4'-dione monoethylene ketal 97%
Bicyclohexane-4,4'-dione monoethylene ketal
Formula: C₁₄H₂₂O₃
MW: 238.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00809704
CAS Number: 56309-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR