2-Amino-9-fluorenone 98%
Supplier: Thermo Scientific Chemicals
H55048-06
H55048-14
H55048-03
AAH55048-06EA
189.99
USD
AAH55048-06
AAH55048-14
AAH55048-03
2-Amino-9-fluorenone 98%
2-Amino-9-fluorenone
Formula:
C₁₃H₉NO MW: 195.22 g/mol Melting Pt: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001157 CAS Number: 3096-57-9 EINECS: 221-445-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...