4-Amino-2-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpyridin-4-amine
, 2-Methyl-4-pyridylamine
, 4-Amino-2-picoline
H55036-06
H55036-MD
H55036-03
AAH55036-MDEA
70.89
USD
AAH55036-MD
AAH55036-06
AAH55036-03
4-Amino-2-methylpyridine 98%
4-Amino-2-methylpyridine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00186509 CAS Number: 18437-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...