(3-Methoxybenzyl)methylamine 97%
|
Danger
|
Synonyms:
1-(3-Methoxyphenyl)-N-methylmethanamine
, 3-Methoxy-N-methylbenzylamine
H54970-06
H54970-03
AAH54970-06EA
200.85
USD
AAH54970-06
AAH54970-03
(3-Methoxybenzyl)methylamine 97%
(3-Methoxybenzyl)methylamine
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 130…133 °C Density: 1.014 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04618244 CAS Number: 41789-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...