4-(2-Chloropyridin-3-yl)pyrimidin-2-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

H54957-MD H54957-03
AAH54957-MDEA 156.51 USD
AAH54957-MD AAH54957-03
4-(2-Chloropyridin-3-yl)pyrimidin-2-amine 97%
4-(2-Chloropyridin-3-yl)pyrimidin-2-amine
Formula: C₉H₇ClN₄
Storage Temperature: Ambient
MDL Number: MFCD18157687
CAS Number: 870221-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR