Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate 97%

Warning

Synonyms: Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate

H54942-06 H54942-MD H54942-03
AAH54942-03EA 61.72 USD
AAH54942-03 AAH54942-06 AAH54942-MD
Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate 97%
Ethyl 4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate
Formula: C₁₂H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD01567252
CAS Number: 39091-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR