2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid 97%

Warning

H54928-MD H54928-03
AAH54928-MDEA 96.68 USD
AAH54928-MD AAH54928-03
2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid 97%
2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid
Formula: C₁₄H₁₂BFN₂O₅
MW: 318.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20265378
CAS Number: 1449131-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR