3,4-Dihydroxythiobenzamide 97%

Warning

H54875-MD H54875-06 H54875-03
AAH54875-MDEA 30.25 USD
AAH54875-MD AAH54875-06 AAH54875-03
3,4-Dihydroxythiobenzamide 97%
3,4-Dihydroxythiobenzamide
Formula: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09047665
CAS Number: 138038-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR