2-Amino-4-(4-cyanophenyl)thiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(2-Amino-1,3-thiazol-4-yl)benzonitrile
H54861-06
H54861-03
H54861-MD
AAH54861-06EA
424.99
USD
AAH54861-06
AAH54861-03
AAH54861-MD
2-Amino-4-(4-cyanophenyl)thiazole 97%
2-Amino-4-(4-cyanophenyl)thiazole
Formula:
C₁₀H₇N₃S MW: 201.25 Storage Temperature: Ambient |
MDL Number:
MFCD03407828 CAS Number: 436151-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...