3,5-Dibromoquinoline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54859-MD
H54859-06
H54859-03
AAH54859-MDEA
34.99
USD
AAH54859-MD
AAH54859-06
AAH54859-03
3,5-Dibromoquinoline 96%
3,5-Dibromoquinoline
Formula:
C₉H₅Br₂N Storage Temperature: Ambient |
MDL Number:
MFCD12024477 CAS Number: 101861-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...