(S)-(-)-3-Chloro-1-phenyl-1-propanol 98+%

Warning

Synonyms: (S)-3-Chloro-1-phenylpropan-1-ol

H54847-06 H54847-03 H54847-MD
AAH54847-06EA 378.01 USD
AAH54847-06 AAH54847-03 AAH54847-MD
(S)-(-)-3-Chloro-1-phenyl-1-propanol 98+%
(S)-(-)-3-Chloro-1-phenyl-1-propanol
Formula: C₉H₁₁ClO
MW: 170.64 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00013309
CAS Number: 100306-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR