Methyl-2-amino-3-bromo-5-methoxybenzoate 96%

Supplier: Thermo Scientific Chemicals
Warning

H54844-MD H54844-03
AAH54844-03EA 339.99 USD
AAH54844-03 AAH54844-MD
Methyl-2-amino-3-bromo-5-methoxybenzoate 96%
Methyl 2-amino-3-bromo-5-methoxybenzoate
Formula: C₉H₁₀BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD20527215
CAS Number: 1378874-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR