Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate 97%

Warning

Synonyms: Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate

H54842-06 H54842-MD H54842-03
AAH54842-06EA 157.62 USD
AAH54842-06 AAH54842-MD AAH54842-03
Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate 97%
Ethyl 4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Formula: C₁₂H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD01936157
CAS Number: 383146-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR