N'-Hydroxy-2-pyrazinecarboximidamide 97%

Warning

Synonyms: Pyrazine-2-amidoxime

H54834-06 H54834-MD H54834-03
AAH54834-03EA 48.09 USD
AAH54834-03 AAH54834-MD AAH54834-06
N'-Hydroxy-2-pyrazinecarboximidamide 97%
N'-Hydroxy-2-pyrazinecarboximidamide
Formula: C₅H₆N₄O
MW: 138.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258397
CAS Number: 51285-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR