2-Methoxy-6-quinolineboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

H54814-03 H54814-MD
AAH54814-MDEA 102.5 USD
AAH54814-MD AAH54814-03
2-Methoxy-6-quinolineboronic acid pinacol ester 96%
2-Methoxy-6-quinolineboronic acid pinacol ester
Formula: C₁₆H₂₀BNO₃
MW: 285.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383956
CAS Number: 1201644-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR