4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%
Synonyms:
4-Fluoro-3-methoxybenzeneboronic acid pinacol ester
, 2-(4-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H54778-MD
H54778-06
H54778-03
AAH54778-06EA
613.35
USD
AAH54778-06
AAH54778-MD
AAH54778-03
4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%
4-Fluoro-3-methoxyphenylboronic acid pinacol ester
Formula:
C₁₃H₁₈BFO₃ MW: 252.09 g/mol Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD11227055 CAS Number: 425378-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...