6-Bromo-4-methyl-3-nitroquinoline 96%

Supplier: Thermo Scientific Chemicals

Warning

H54769-MD H54769-03
AAH54769-MDEA 140.73 USD
AAH54769-MD AAH54769-03
6-Bromo-4-methyl-3-nitroquinoline 96%
6-Bromo-4-methyl-3-nitroquinoline
Formula: C₁₀H₇BrN₂O₂
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD12761249
CAS Number: 1185310-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR