3-(3-Methylphenyl)-5-phenyl-1,2,4-oxadiazole 97%

Warning

H54748-MD H54748-06 H54748-03
AAH54748-06EA 374.07 USD
AAH54748-06 AAH54748-03 AAH54748-MD
3-(3-Methylphenyl)-5-phenyl-1,2,4-oxadiazole 97%
3-(3-Methylphenyl)-5-phenyl-1,2,4-oxadiazole
Formula: C₁₅H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD03884617
CAS Number: 111249-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR