2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid 97%

Warning

H54713-06 H54713-03 H54713-MD
AAH54713-03EA 79.64 USD
AAH54713-03 AAH54713-06 AAH54713-MD
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid 97%
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid
Formula: C₁₂H₁₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD01316979
CAS Number: 23353-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR