5-Bromo-2-methylpyridin-3-amine 96%
|
Warning
|
Synonyms:
3-Amino-5-bromo-2-methylpyridine
H54665-MD
H54665-03
AAH54665-03EA
270.91
USD
AAH54665-03
AAH54665-MD
5-Bromo-2-methylpyridin-3-amine 96%
5-Bromo-2-methylpyridin-3-amine
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Melting Pt: 108…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD09031418 CAS Number: 914358-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...