3-(4-Methylpiperidin-1-ylsulfonyl)phenylboronic acid 97%

Warning

Synonyms: 3-[(4-Methyl-1-piperidinyl)sulphonyl]benzeneboronic acid

H54664-MD H54664-03
AAH54664-MDEA 139.41 USD
AAH54664-MD AAH54664-03
3-(4-Methylpiperidin-1-ylsulfonyl)phenylboronic acid 97%
3-(4-Methylpiperidin-1-ylsulphonyl)phenylboronic acid
Formula: C₁₂H₁₈BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD20265316
CAS Number: 1449132-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR