Indole-2-carbaldehyd 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indole-2-carboxaldehyde
, 1H-Indole-2-carbaldehyde
H54638-MD
H54638-03
H54638-06
AAH54638-MDEA
44.89
USD
AAH54638-MD
AAH54638-06
AAH54638-03
Indole-2-carbaldehyd 97%
Indole-2-carbaldehyd
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD03001425 CAS Number: 19005-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...