3,4-Difluorobenzamidoxime 97%
|
Warning
|
H54636-06
H54636-MD
H54636-03
AAH54636-MDEA
34.76
USD
AAH54636-MD
AAH54636-06
AAH54636-03
3,4-Difluorobenzamidoxime 97%
3,4-Difluorobenzamidoxime
Formula:
C₇H₆F₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD06660194 CAS Number: 500024-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...