3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid 97%
|
Warning
|
H54634-03
H54634-MD
AAH54634-MDEA
125.43
USD
AAH54634-MD
AAH54634-03
3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid 97%
3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid
Formula:
C₁₁H₁₃BFNO₃ MW: 237.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20265305 CAS Number: 1264616-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...