2-(Methylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 2-(Methylcarbamoyl)benzeneboronic acid

H54628-MD H54628-03
AAH54628-03EA 397.38 USD
AAH54628-03 AAH54628-MD
2-(Methylcarbamoyl)phenylboronic acid 97%
2-(Methylcarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD22190724
CAS Number: 874459-85-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR