N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate ≥98%

Warning

Synonyms: 4-(N-Maleimidomethyl)cyclohexanecarboxylic Acid N-Succinimidyl EsterSMCC

H54627-MC H54627-MB
AAH54627-MBEA 76.46 USD
AAH54627-MB AAH54627-MC
N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate ≥98%
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Formula: C₁₆H₁₈N₂O₆
MW: 334.33 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009634
CAS Number: 64987-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR