2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid 97%
|
Warning
|
H54606-03
H54606-06
H54606-MD
AAH54606-03EA
88.41
USD
AAH54606-03
AAH54606-MD
AAH54606-06
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid 97%
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid
Formula:
C₁₁H₉NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD03791177 CAS Number: 57677-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...