4,5-Dicyano-2-(2-methylphenyl)imidazole 97%
|
Warning
|
H54582-03
H54582-06
H54582-MD
AAH54582-MDEA
31.89
USD
AAH54582-MD
AAH54582-06
AAH54582-03
4,5-Dicyano-2-(2-methylphenyl)imidazole 97%
4,5-Dicyano-2-(2-methylphenyl)imidazole
Formula:
C₁₂H₈N₄ Storage Temperature: Ambient |
MDL Number:
MFCD20265294 CAS Number: 63857-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...