2-Chloro-5-(cyclopropylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid

H54576-MD H54576-03
AAH54576-03EA 579.42 USD
AAH54576-03 AAH54576-MD
2-Chloro-5-(cyclopropylcarbamoyl)phenylboronic acid 97%
2-Chloro-5-(cyclopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₁BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD22125077
CAS Number: 1449132-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR