2-Chloro-5-(2,3-dichlorophenylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 2-Chloro-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid

H54574-MD H54574-03
AAH54574-MDEA 194.25 USD
AAH54574-MD AAH54574-03
2-Chloro-5-(2,3-dichlorophenylcarbamoyl)phenylboronic acid 97%
2-Chloro-5-(2,3-dichlorophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₉BCl₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD27996215
CAS Number: 1449144-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR