3,4-Difluorothiobenzamide 97%
|
Warning
|
H54564-06
H54564-03
H54564-MD
AAH54564-MDEA
27.79
USD
AAH54564-MD
AAH54564-06
AAH54564-03
3,4-Difluorothiobenzamide 97%
3,4-Difluorothiobenzamide
Formula:
C₇H₅F₂NS Storage Temperature: Ambient |
MDL Number:
MFCD04036083 CAS Number: 317319-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...