4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54552-06
H54552-03
H54552-MD
AAH54552-MDEA
68.27
USD
AAH54552-MD
AAH54552-06
AAH54552-03
4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
4-Chloro-5,6-dimethylpyrimidin-2-ylamine
Formula:
C₆H₈ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD00462111 CAS Number: 14394-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...