Methyl picolinate 99%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Methyl pyridine-2-carboxylate

H54530-22 H54530-14
AAH54530-14EA 118 USD
AAH54530-14 AAH54530-22
Methyl picolinate 99%
Methyl picolinate
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 95 °C/1 mm
Melting Pt: 19 °C
Density: 1.137
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038038
CAS Number: 2459-07-6
EINECS: 219-545-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR