2-Fluoro-5-(2-fluorobenzylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 2-Fluoro-5-(2-fluorobenzylcarbamoyl)benzeneboronic acid

H54510-03 H54510-MD
AAH54510-03EA 436.23 USD
AAH54510-03 AAH54510-MD
2-Fluoro-5-(2-fluorobenzylcarbamoyl)phenylboronic acid 97%
2-Fluoro-5-(2-fluorobenzylcarbamoyl)phenylboronic acid
Formula: C₁₄H₁₂BF₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD20040208
CAS Number: 1449144-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR