2-Bromo-4-methylpyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54503-14
H54503-06
H54503-03
AAH54503-06EA
260.99
USD
AAH54503-06
AAH54503-14
AAH54503-03
2-Bromo-4-methylpyrimidine 97%
2-Bromo-4-methylpyrimidine
Formula:
C₅H₅ClN₂ Melting Pt: 45…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD04037883 CAS Number: 130645-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...