2-Chloro-5-(2,5-difluorophenylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 2-Chloro-5-(2,5-difluorophenylcarbamoyl)benzeneboronic acid

H54492-MD H54492-03
AAH54492-MDEA 194.25 USD
AAH54492-MD AAH54492-03
2-Chloro-5-(2,5-difluorophenylcarbamoyl)phenylboronic acid 97%
2-Chloro-5-(2,5-difluorophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₉BClF₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD20040200
CAS Number: 1449132-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR