1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
, 4-(6-Nitropyridin-3-yl)piperazine, N1-BOC protected
H54465-06
H54465-MD
H54465-03
AAH54465-06EA
521.99
USD
AAH54465-06
AAH54465-03
AAH54465-MD
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
1-Boc-4-(6-nitro-3-pyridyl)piperazine
Formula:
C₁₄H₂₀N₄O₄ MW: 308.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11849291 CAS Number: 571189-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...