Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-pyrimidineboronic acid pinacol ester ≥96%

Supplier: Alfa Aesar
Warning

H54451-06 H54451-MD H54451-03
AAH54451-06EA 487.19 USD InStock
AAH54451-06 AAH54451-MD AAH54451-03
2-Amino-5-pyrimidineboronic acid pinacol ester ≥96%
2-Amino-5-pyrimidineboronic acid pinacol ester


Formula: C₁₀H₁₆BN₃O₂
MW: 221,06
Melting Pt: 206…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06795685
CAS Number: 402960-38-7
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty