4,6-Dichloropyrimidine-5-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54449-14
H54449-06
H54449-03
AAH54449-14EA
608.99
USD
AAH54449-14
AAH54449-03
AAH54449-06
4,6-Dichloropyrimidine-5-carbaldehyde 97%
4,6-Dichloropyrimidine-5-carbaldehyde
Formula:
C₅H₂Cl₂N₂O MW: 175.99 g/mol Melting Pt: 66…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD02257701 CAS Number: 5305-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...