β-Lactose mixture of stereoisomers ∼70% beta & ∼30% alpha
Supplier: Thermo Scientific Chemicals
Synonyms:
beta-D-Lactose
, β-D-Gal-(1→4)-β-D-Glc
H54447-18
H54447-30
H54447-A1
AAH54447-30EA
159.23
USD
AAH54447-30
AAH54447-A1
AAH54447-18
β-Lactose mixture of stereoisomers ∼70% beta & ∼30% alpha
β-Lactose
Formula:
C₁₂H₂₂O₁₁ MW: 342.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064521 CAS Number: 5965-66-2 EINECS: 227-751-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...