Indole-3-thiocarboxamide 97%

Warning

H54438-MD H54438-03
AAH54438-03EA 229.77 USD
AAH54438-03 AAH54438-MD
Indole-3-thiocarboxamide 97%
Indole-3-thiocarboxamide
Formula: C₉H₈N₂S
MW: 176.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01993552
CAS Number: 59108-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR