2-Chloro-5-(2,5-dichlorophenylcarbamoyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid
H54433-03
H54433-MD
AAH54433-03EA
579.42
USD
AAH54433-03
AAH54433-MD
2-Chloro-5-(2,5-dichlorophenylcarbamoyl)phenylboronic acid 97%
2-Chloro-5-(2,5-dichlorophenylcarbamoyl)phenylboronic acid
Formula:
C₁₃H₉BCl₃NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22125068 CAS Number: 1449132-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...