2-Chloro-5-(cyclohexylcarbamoyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
2-Chloro-5-(cyclohexylcarbamoyl)benzeneboronic acid
H54425-MD
H54425-03
AAH54425-MDEA
198.69
USD
AAH54425-MD
AAH54425-03
2-Chloro-5-(cyclohexylcarbamoyl)phenylboronic acid 97%
2-Chloro-5-(cyclohexylcarbamoyl)phenylboronic acid
Formula:
C₁₃H₁₇BClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22125067 CAS Number: 1449132-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...