5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%

Warning

Synonyms: 4-(5-Methyl-1,2,4-oxadiazol-3-yl)pyridine

H54416-MD H54416-03 H54416-06
AAH54416-06EA 304.14 USD
AAH54416-06 AAH54416-MD AAH54416-03
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole
Formula: C₈H₇N₃O
MW: 161.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464110
CAS Number: 10350-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR