N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl EsterDABCYL N-Succinimidyl Ester

H54402-MB H54402-MC
AAH54402-MBEA 62.03 USD
AAH54402-MB AAH54402-MC
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
Formula: C₁₉H₁₈N₄O₄
MW: 366.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00467415
CAS Number: 146998-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR