2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
, 4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester
H54375-03
H54375-MD
AAH54375-MDEA
152.99
USD
AAH54375-MD
AAH54375-03
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester 96%
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester
Formula:
C₁₁H₁₈BN₃O₂ MW: 235.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18072554 CAS Number: 944401-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...