4-Acetoxybenzonitrile 97%

Warning

Synonyms: 4-Cyanophenyl acetate

H54358-06 H54358-03
AAH54358-06EA 87.46 USD
AAH54358-06 AAH54358-03
4-Acetoxybenzonitrile 97%
4-Acetoxybenzonitrile
Formula: C₉H₇NO₂
MW: 161.16
Storage Temperature: Ambient
EINECS: 235-893-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR