2-(2-Thienyl)-4-thiazolecarboxylic acid 97%

Warning

H54347-06 H54347-MD H54347-03
AAH54347-06EA 473.88 USD
AAH54347-06 AAH54347-MD AAH54347-03
2-(2-Thienyl)-4-thiazolecarboxylic acid 97%
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formula: C₈H₅NO₂S₂
MW: 211.25
Storage Temperature: Ambient
MDL Number: MFCD01936014
CAS Number: 24044-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR