2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester 96%
|
Warning
|
H54341-03
H54341-MD
AAH54341-03EA
266.23
USD
AAH54341-03
AAH54341-MD
2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester 96%
2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester
Formula:
C₁₈H₂₃BN₂O₃ MW: 326.20 g/mol Melting Pt: 135…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD13191381 CAS Number: 756520-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...